This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01261293_m1
See other CREBL2 GE Assays ›
Gene Symbol:
Crebl2
Gene Name:
cAMP responsive element binding protein-like 2
Gene Aliases:
AI046348, B230205M03
Chromosome Location:
Chr.6: 134828686 - 134858931 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01261293_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Crebl2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177687.3 NP_808355.1 1 - 2 247 61
GenBank mRNA AK045467.1 - 1 - 2 248 61
BC109358.1 - 1 - 2 119 40057562 61
BC109359.1 - 1 - 2 135 40057564 61

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK045467.1, BC109358.1, BC109359.1, NM_177687.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Crebl2

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
cell differentiation
positive regulation of peptidyl-serine phosphorylation
positive regulation of fat cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of glucose import
positive regulation of lipid biosynthetic process
protein stabilization
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0