This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01261100_m1
See other ACSL5 GE Assays ›
Gene Symbol:
Acsl5
Gene Name:
acyl-CoA synthetase long-chain family member 5
Gene Aliases:
1700030F05Rik, ACS2, ACS5, Facl5
Chromosome Location:
Chr.19: 55251862 - 55296628 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01261100_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acsl5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027976.2 NP_082252.1 9 - 10 1620 66
GenBank mRNA AK034942.1 - 1620 66
AK145894.1 - 9 - 10 1281 66
AK150542.1 - 9 - 10 1286 66
AK150663.1 - 9 - 10 1286 66
AK151378.1 - 9 - 10 1075 66
AK161024.1 - 9 - 10 973 66
AK206020.1 - 2 - 3 87 66
AK219510.1 - 2 - 3 87 66
BC031544.1 - 9 - 10 1054 3987201 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000776.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Acsl5

Panther Classification:

Molecular Function -

ligase

Gene Ontology Categories:

Function(s) Process(es)

long-chain fatty acid metabolic process
lipid metabolic process
fatty acid metabolic process
metabolic process
phospholipid biosynthetic process
positive regulation of plasma membrane long-chain fatty acid transport
positive regulation of triglyceride biosynthetic process
fatty acid transport
positive regulation of fatty acid beta-oxidation
regulation of extrinsic apoptotic signaling pathway
nucleotide binding
catalytic activity
long-chain fatty acid-CoA ligase activity
ATP binding
ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900006833.mm.1 NM_027976.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900006833.mm.1 NM_027976.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900006833.mm.1 TC1900000776.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0