This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01259581_m1
See other 4930479D17RIK GE Assays ›
Gene Symbol:
4930479D17Rik
Gene Name:
RIKEN cDNA 4930479D17 gene
Gene Aliases:
-
Chromosome Location:
Chr.6: 146657218 - 146665403 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01259581_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930479D17Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_046277.1 2 - 3 550 79
GenBank mRNA AK015591.1 - 2 - 3 551 79

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream mediator complex subunit 21 Med21 0610007L03Rik,
AI449604,
D19234,
D6Ertd782e,
Srb7,
Surb7
-13.5kb Sense Chr. 6 - 146642579 - 146650600 on NCBI Build GRCm38
Downstream serine/threonine kinase 38 like Stk38l 4930473A22Rik,
B230328I19,
Ndr2,
Ndr54
59.5kb Sense Chr. 6 - 146724930 - 146778814 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904344 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 149011 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 602179 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 90284 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159042 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119219 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24929868 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24929933 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24929867 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24929923 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05428250 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24929946 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24929916 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24929875 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24929939 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24929948 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24929913 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05118291 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24929938 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24929944 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24929921 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24929870 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05055764 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24929940 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24929915 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24929866 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24929945 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05248146 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24929942 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24929931 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24929914 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05055770 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24929918 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24929871 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24929927 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600013760.mm.1 NR_046277.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013760.mm.1 NR_046277.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013760.mm.1 TC0600001735.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600013761.mm.1 NR_046277.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013761.mm.1 NR_046277.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013761.mm.1 TC0600001735.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600013756.mm.1 NR_046277.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013756.mm.1 NR_046277.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013756.mm.1 TC0600001735.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600013757.mm.1 NR_046277.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013757.mm.1 NR_046277.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013757.mm.1 TC0600001735.mm.1 MTA TC
Murine Genome U74v2 Set 140446_at - -
Murine Genome U74v2 Set 140446_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 427250 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 104060427 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0