This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01259317_m1
See other PRPF4 GE Assays ›
Gene Symbol:
Prpf4
Gene Name:
PRP4 pre-mRNA processing factor 4 homolog (yeast)
Gene Aliases:
1600015H11Rik, AI874830, AW047464, bN189G18.1
Chromosome Location:
Chr.4: 62408767 - 62426990 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01259317_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prpf4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027297.3 NP_081573.1 8 - 9 896 68
GenBank mRNA AK005464.1 - 8 - 9 875 68
AK031219.1 - 882 68
AK085174.1 - 8 - 9 860 68
AK089838.1 - 8 - 9 848 68
AK090114.1 - 8 - 9 873 68
AK166912.1 - 8 - 9 882 68
AK167216.1 - 8 - 9 874 68
AK169532.1 - 8 - 9 859 68
BC051639.1 - 4 - 5 310 68
BC106089.1 - 8 - 9 858 30715403 68
BC125529.1 - 8 - 9 847 40130044 68
BC125531.1 - 8 - 9 847 40130046 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400000700.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Prpf4

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
mRNA processing
RNA splicing
molecular_function
U6 snRNA binding
U4 snRNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400004917.mm.1 NM_027297.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400004917.mm.1 NM_027297.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400004917.mm.1 TC0400000700.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0