This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01259019_g1
See other SLC4A3 GE Assays ›
Gene Symbol:
Slc4a3
Gene Name:
solute carrier family 4 (anion exchanger), member 3
Gene Aliases:
A930038D23Rik, Ae3
Chromosome Location:
Chr.1: 75546041 - 75559431 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01259019_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc4a3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009208.3 NP_033234.2 8 - 9 1350 73
GenBank mRNA BC066086.1 - 9 - 10 1209 73
BC080271.1 - 8 - 9 1345 6831942 73
M28383.1 - 8 - 9 1320 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000665.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc4a3

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
anion transport
bicarbonate transport
regulation of intracellular pH
anion transmembrane transport
chloride transmembrane transport
transporter activity
inorganic anion exchanger activity
anion transmembrane transporter activity
antiporter activity
anion:anion antiporter activity
enzyme binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100005757.mm.1 NM_009208.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100005757.mm.1 NM_009208.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100005757.mm.1 TC0100000665.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0