This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01258146_gH
See other CAMK2A GE Assays ›
Gene Symbol:
Camk2a
Gene Name:
calcium/calmodulin-dependent protein kinase II alpha
Gene Aliases:
CaMKII, R74975, mKIAA0968
Chromosome Location:
Chr.18: 60925580 - 60988152 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01258146_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Camk2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177407.4 NP_803126.1 10 - 11 969 67
GenBank mRNA AK083245.1 - 10 - 11 970 67
AK122410.2 - 10 - 11 965 67
BC031745.1 - 10 - 11 950 4506005 67
X14836.1 - 10 - 11 953 67

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Camk2a

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
response to ischemia
protein phosphorylation
calcium ion transport
positive regulation of cardiac muscle cell apoptotic process
phosphorylation
peptidyl-serine phosphorylation
ionotropic glutamate receptor signaling pathway
angiotensin-activated signaling pathway
protein autophosphorylation
regulation of neurotransmitter secretion
regulation of neuronal synaptic plasticity
positive regulation of NF-kappaB transcription factor activity
positive regulation of calcium ion transport
regulation of mitochondrial membrane permeability involved in apoptotic process
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
protein binding
calmodulin binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
GTPase activating protein binding
glutamate receptor binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800004456.mm.1 NM_177407.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004456.mm.1 NM_177407.4 Refseq
Affymetrix Mouse Clariom S Array PSR1800004456.mm.1 TC1800000620.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header