This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01258019_m1
See other 4930552P12RIK GE Assays ›
Gene Symbol:
4930552P12Rik
Gene Name:
RIKEN cDNA 4930552P12 gene
Gene Aliases:
-
Chromosome Location:
Chr.19: 55640589 - 55702204 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01258019_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930552P12Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045318.1 3 - 4 533 65
GenBank mRNA AK019762.1 - 3 - 4 534 65
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY715703.1 - 3 - 4 534 Chr. 19 - 55642420 - 55702204 [-] on Build GRCm38 65

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream vesicle transport through interaction with t-SNAREs 1A Vti1a 1110014F16Rik,
1110018K19Rik,
4921537J05Rik,
MVti1,
MVti1a,
Vti1,
Vti1-rp2
-13.1kb Anti-sense Chr. 19 - 55316057 - 55627461 on NCBI Build GRCm38
Downstream transcription factor 7 like 2,
T cell specific,
HMG box
Tcf7l2 TCF4B,
TCF4E,
Tcf-4,
Tcf4,
mTcf-4B,
mTcf-4E
98.2kb Anti-sense Chr. 19 - 55741810 - 55933661 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900001699.mm.1
This assay targets the same location as RTA transcript cluster(s) TC0100003843.rn

Set Membership:

Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2387865 AK019762.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 499523 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 700580 AK034246.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 405637 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 308879 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 283209 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 585282 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 664195 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 546181 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 953612 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 402173 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 520316 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 91011 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 254969 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 68346 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 225646 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 329963 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 317531 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431234 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 896973 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 969741 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 868520 AK047890.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 236269 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 365343 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 963779 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 77436 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 12493 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 88056 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 79953 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64186 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146736 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03622792 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146736 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09207739 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09207817 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09207800 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136117 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146736 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195896 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195966 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195613 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195907 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195979 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195734 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32196003 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14352795 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195596 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195588 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195745 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195688 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195747 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195911 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195935 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14286201 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195604 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195944 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195981 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195994 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195883 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195737 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14352785 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195589 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195778 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14420280 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195941 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195698 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195730 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195988 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195706 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195905 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195738 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195771 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195936 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14470174 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14462185 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14352800 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195774 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195885 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195995 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32196007 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195984 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195593 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14255271 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195726 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195764 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195964 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195938 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195772 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195913 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32196012 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195906 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195893 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195607 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195765 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14433966 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14433984 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195997 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14495263 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195615 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195924 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32196010 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14470172 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195970 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195601 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195735 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195900 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195931 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195611 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14230338 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195954 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195890 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195918 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195600 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14412610 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14303228 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195889 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195586 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14264898 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195898 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195760 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195590 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32196001 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195610 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195902 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14188557 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195592 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195749 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195729 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195699 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195891 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14366307 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14352778 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195960 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195597 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195694 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195703 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195933 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14189811 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195758 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195969 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14366303 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14255268 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195929 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195608 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14329334 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195606 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195999 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32196005 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14188552 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195990 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195940 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195917 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14507760 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195953 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195701 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195955 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195602 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195696 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195768 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195599 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195986 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195690 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32195915 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1460507_PM_at - -
Mouse Genome 430 2.0 Array 1460507_PM_at - -
Mouse Genome 430 2.0 Array 1460507_at - -
Affymetrix Mouse Clariom S Array PSR1900014913.mm.1 NR_045318.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900014913.mm.1 NR_045318.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900014913.mm.1 TC1900001699.mm.1 MTA TC
Mouse Expression Set 430 1460507_at - -
Mouse Expression Set 430 1460507_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1460507_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1460507_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 602484 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 100730162 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0