This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01257824_m1
See other PPP2R3C GE Assays ›
Gene Symbol:
Ppp2r3c
Gene Name:
protein phosphatase 2, regulatory subunit B'', gamma
Gene Aliases:
4930511A21Rik, 5730412A08Rik, G4-1, G5pr
Chromosome Location:
Chr.12: 55280809 - 55303005 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01257824_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp2r3c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021529.3 NP_067504.2 11 - 12 1201 71
GenBank mRNA AJ238247.2 - 10 - 11 1028 71
AK015746.1 - 10 - 11 1030 71
AK028063.1 - 11 - 12 1166 71
AK028254.1 - 10 - 11 1120 71
AK033405.1 - 11 - 12 1200 71
AK040589.1 - 10 - 11 1029 71
AK087572.1 - 10 - 11 1024 71
AK088240.1 - 10 - 11 1110 71
AK168298.1 - 11 - 12 1203 71
BC024754.1 - 11 - 12 1164 5375405 71
BC138295.1 - 10 - 11 997 8861314 71
BC138296.1 - 10 - 11 997 8861315 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200001763.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp2r3c

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
B cell homeostasis
regulation of antimicrobial humoral response
cortical cytoskeleton organization
activation of protein kinase activity
T cell homeostasis
positive regulation of B cell differentiation
spleen development
regulation of B cell activation
regulation of mitochondrial depolarization
protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200012153.mm.1 NM_021529.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200012153.mm.1 NM_021529.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200012153.mm.1 TC1200001763.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200012154.mm.1 NM_021529.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200012154.mm.1 NM_021529.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200012154.mm.1 TC1200001763.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0