Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400023785.mm.1 |
NM_001113209.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023785.mm.1 |
NM_001113210.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023785.mm.1 |
NM_001286127.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023785.mm.1 |
NM_001286131.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023785.mm.1 |
NM_008687.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023785.mm.1 |
TC0400002929.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400023791.mm.1 |
NM_001113209.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023791.mm.1 |
NM_001113210.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023791.mm.1 |
NM_001286127.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023791.mm.1 |
NM_001286131.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023791.mm.1 |
NM_008687.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023791.mm.1 |
TC0400002929.mm.1 |
MTA TC |