This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01257278_m1
See other STAT1 GE Assays ›
Gene Symbol:
Stat1
Gene Name:
signal transducer and activator of transcription 1
Gene Aliases:
2010005J02Rik, AA408197
Chromosome Location:
Chr.1: 52119438 - 52161865 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01257278_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stat1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001205313.1 NP_001192242.1 11 - 12 1332 160
NM_001205314.1 NP_001192243.1 11 - 12 1368 160
NM_009283.4 NP_033309.3 11 - 12 1383 160
GenBank mRNA AK018544.1 - 11 - 12 1334 160
AK039458.1 - 11 - 12 1384 160
AK046517.1 - 11 - 12 1332 160
AK082706.1 - 11 - 12 1365 160
AK084855.1 - 11 - 12 1317 160
AK157263.1 - 11 - 12 1232 160
AK159887.1 - 11 - 12 1367 160
BC004808.1 - 11 - 12 1319 3587831 160

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000375.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stat1

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of endothelial cell proliferation
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
activation of cysteine-type endopeptidase activity involved in apoptotic process
signal transduction
JAK-STAT cascade
response to nutrient
blood circulation
positive regulation of cell proliferation
response to mechanical stimulus
response to bacterium
negative regulation of angiogenesis
cytokine-mediated signaling pathway
lipopolysaccharide-mediated signaling pathway
response to lipopolysaccharide
cellular response to insulin stimulus
tumor necrosis factor-mediated signaling pathway
response to cytokine
negative regulation of macrophage fusion
cellular response to interferon-beta
response to drug
negative regulation of I-kappaB kinase/NF-kappaB signaling
response to exogenous dsRNA
response to peptide hormone
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation by virus of viral protein levels in host cell
positive regulation of smooth muscle cell proliferation
response to cAMP
interferon-gamma-mediated signaling pathway
type I interferon signaling pathway
renal tubule development
cellular response to lipopolysaccharide
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
transcription factor activity, RNA polymerase II core promoter sequence-specific
DNA binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
signal transducer activity
tumor necrosis factor receptor binding
protein binding
enzyme binding
CCR5 chemokine receptor binding
identical protein binding
protein homodimerization activity
sequence-specific DNA binding
protein phosphatase 2A binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0