This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01256386_g1
See other BIVM GE Assays ›
Gene Symbol:
Bivm
Gene Name:
basic, immunoglobulin-like variable motif containing
Gene Aliases:
AI854267
Chromosome Location:
Chr.1: 44118888 - 44144771 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01256386_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bivm
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144558.4 NP_653141.3 4 - 5 1132 129
GenBank mRNA AF411386.1 - 5 - 6 1135 129
AK032375.1 - 4 - 5 1092 129
AK034329.1 - 4 - 5 1094 129
BC082584.1 - 4 - 5 938 30612339 129

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000333.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bivm

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100002475.mm.1 NM_144558.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100002475.mm.1 NM_144558.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100002475.mm.1 TC0100000333.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0