Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100002504.mm.1 |
NM_011729.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002504.mm.1 |
NM_011729.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002504.mm.1 |
TC0100000334.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100002505.mm.1 |
NM_011729.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002505.mm.1 |
NM_011729.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002505.mm.1 |
TC0100000334.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100002502.mm.1 |
NM_011729.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002502.mm.1 |
NM_011729.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002502.mm.1 |
TC0100000334.mm.1 |
MTA TC |