This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01256326_m1
See other ERCC5 GE Assays ›
Gene Symbol:
Ercc5
Gene Name:
excision repair cross-complementing rodent repair deficiency, complementation group 5
Gene Aliases:
Xpg
Chromosome Location:
Chr.1: 44147578 - 44181260 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01256326_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ercc5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011729.2 NP_035859.2 4 - 5 789 69
GenBank mRNA AK038822.1 - 4 - 5 618 69
AK137563.1 - 4 - 5 621 69
AK140825.1 - 4 - 5 618 69
AK160645.1 - 2 - 3 161 69
AK164205.1 - 4 - 5 607 69
BC033413.1 - 4 - 5 509 69
BC060659.1 - 4 - 5 588 69
BC092235.1 - 4 - 5 580 69
BC141070.1 - 4 - 5 619 9055682 69
D16306.1 - 4 - 5 484 69
U40796.1 - 4 - 5 685 69

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ercc5

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
transcription-coupled nucleotide-excision repair
nucleotide-excision repair
nucleotide-excision repair, DNA incision, 3'-to lesion
cellular response to DNA damage stimulus
determination of adult lifespan
response to UV
UV protection
response to UV-C
multicellular organism growth
negative regulation of apoptotic process
bubble DNA binding
DNA binding
double-stranded DNA binding
single-stranded DNA binding
catalytic activity
nuclease activity
endonuclease activity
endodeoxyribonuclease activity
hydrolase activity
hydrolase activity, acting on ester bonds
phosphoric ester hydrolase activity
protein homodimerization activity
T/G mismatch-specific endonuclease activity
retroviral 3' processing activity
metal ion binding
protein N-terminus binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0