This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01255577_m1
See other CFLAR GE Assays ›
Gene Symbol:
Cflar
Gene Name:
CASP8 and FADD-like apoptosis regulator
Gene Aliases:
2310024N18Rik, A430105C05Rik, CLARP, Cash, Casper, ENSMUSG00000072980, FLAME, FLAME-1, Flip, Gm9845, I-FLICE, MRIT, c-Flip
Chromosome Location:
Chr.1: 58711310 - 58759209 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01255577_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cflar
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289704.2 NP_001276633.1 6 - 7 903 69
NM_001293804.1 NP_001280733.1 6 - 7 886 69
NM_001293805.1 NP_001280734.1 4 - 5 379 69
NM_207653.5 NP_997536.1 6 - 7 1175 69
GenBank mRNA AK152515.1 - 6 - 7 870 69
BC029223.1 - 6 - 7 1144 4460554 69
U97076.1 - 6 - 7 841 69
Y14041.1 - 5 - 6 753 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000439.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cflar

Panther Classification:

Molecular Function -

protease

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of peptidase activity
negative regulation of apoptotic process
positive regulation of catalytic activity
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of necroptotic process
negative regulation of necroptotic process
apoptotic signaling pathway
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of extrinsic apoptotic signaling pathway
protease binding
death receptor binding
enzyme activator activity
peptidase activator activity
protein complex binding
death effector domain binding
protein heterodimerization activity
cysteine-type endopeptidase activity involved in apoptotic signaling pathway

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100003583.mm.1 NM_001289704.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100003583.mm.1 NM_207653.5 Refseq
Affymetrix Mouse Clariom S Array PSR0100003583.mm.1 TC0100000439.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0