This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01255576_m1
See other CFLAR GE Assays ›
Gene Symbol:
Cflar
Gene Name:
CASP8 and FADD-like apoptosis regulator
Gene Aliases:
2310024N18Rik, A430105C05Rik, CLARP, Cash, Casper, ENSMUSG00000072980, FLAME, FLAME-1, Flip, Gm9845, I-FLICE, MRIT, c-Flip
Chromosome Location:
Chr.1: 58711310 - 58759209 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01255576_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cflar
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289704.2 NP_001276633.1 5 - 6 846 83
NM_001293804.1 NP_001280733.1 5 - 6 829 83
NM_001293805.1 NP_001280734.1 3 - 4 322 83
NM_207653.5 NP_997536.1 5 - 6 1118 83
GenBank mRNA AK152515.1 - 5 - 6 813 83
BC029223.1 - 5 - 6 1087 4460554 83
U97076.1 - 5 - 6 784 83
Y14041.1 - 4 - 5 696 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000439.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protease Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cflar

Panther Classification:

Molecular Function -

protease

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of peptidase activity
negative regulation of apoptotic process
positive regulation of catalytic activity
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of necroptotic process
negative regulation of necroptotic process
apoptotic signaling pathway
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of extrinsic apoptotic signaling pathway
protease binding
death receptor binding
enzyme activator activity
peptidase activator activity
protein complex binding
death effector domain binding
protein heterodimerization activity
cysteine-type endopeptidase activity involved in apoptotic signaling pathway

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100003554.mm.1 NM_001289704.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100003554.mm.1 TC0100000439.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100003576.mm.1 NM_001289704.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100003576.mm.1 NM_009805.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100003576.mm.1 NM_009805.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100003576.mm.1 NM_207653.5 Refseq
Affymetrix Mouse Clariom S Array PSR0100003576.mm.1 TC0100000439.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100003577.mm.1 NM_001289704.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100003577.mm.1 NM_009805.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100003577.mm.1 NM_009805.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100003577.mm.1 NM_207653.5 Refseq
Affymetrix Mouse Clariom S Array PSR0100003577.mm.1 TC0100000439.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100003582.mm.1 NM_001289704.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100003582.mm.1 NM_207653.5 Refseq
Affymetrix Mouse Clariom S Array PSR0100003582.mm.1 TC0100000439.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100003574.mm.1 NM_001289704.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100003574.mm.1 NM_009805.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100003574.mm.1 NM_009805.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100003574.mm.1 NM_207653.5 Refseq
Affymetrix Mouse Clariom S Array PSR0100003574.mm.1 TC0100000439.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100003578.mm.1 NM_001289704.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100003578.mm.1 NM_009805.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100003578.mm.1 NM_009805.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100003578.mm.1 NM_207653.5 Refseq
Affymetrix Mouse Clariom S Array PSR0100003578.mm.1 TC0100000439.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0