This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01255401_m1
See other AOX2 GE Assays ›
Gene Symbol:
Aox2
Gene Name:
aldehyde oxidase 2
Gene Aliases:
Aoh3, Aox3l1
Chromosome Location:
Chr.1: 58276141 - 58379264 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01255401_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aox2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001008419.2 NP_001008419.1 13 - 14 1362 107
GenBank mRNA AY187018.2 - 13 - 14 1362 107
AY665589.1 - 13 - 14 1269 107
BC150826.1 - 13 - 14 1289 9087737 107

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000431.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aox2

Gene Ontology Categories:

Function(s) Process(es)

biological_process
oxidation-reduction process
molecular_function
catalytic activity
aldehyde oxidase activity
iron ion binding
UDP-N-acetylmuramate dehydrogenase activity
electron carrier activity
oxidoreductase activity
oxidoreductase activity, acting on CH-OH group of donors
metal ion binding
flavin adenine dinucleotide binding
NAD binding
iron-sulfur cluster binding
2 iron, 2 sulfur cluster binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100003443.mm.1 NM_001008419.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100003443.mm.1 NM_001008419.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100003443.mm.1 TC0100000431.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100003442.mm.1 NM_001008419.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100003442.mm.1 NM_001008419.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100003442.mm.1 TC0100000431.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0