This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01255400_m1
See other AOX2 GE Assays ›
Gene Symbol:
Aox2
Gene Name:
aldehyde oxidase 2
Gene Aliases:
Aoh3, Aox3l1
Chromosome Location:
Chr.1: 58276141 - 58379264 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01255400_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aox2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001008419.2 NP_001008419.1 12 - 13 1252 75
GenBank mRNA AY187018.2 - 12 - 13 1252 75
AY665589.1 - 12 - 13 1159 75
BC150826.1 - 12 - 13 1179 9087737 75

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aox2

Gene Ontology Categories:

Function(s) Process(es)

biological_process
oxidation-reduction process
molecular_function
catalytic activity
aldehyde oxidase activity
iron ion binding
UDP-N-acetylmuramate dehydrogenase activity
electron carrier activity
oxidoreductase activity
oxidoreductase activity, acting on CH-OH group of donors
metal ion binding
flavin adenine dinucleotide binding
NAD binding
iron-sulfur cluster binding
2 iron, 2 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header