This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01254543_m1
See other INO80D GE Assays ›
Gene Symbol:
Ino80d
Gene Name:
INO80 complex subunit D
Gene Aliases:
7330405I11, A430093A21Rik, BC010584, OTTMUSG00000002594
Chromosome Location:
Chr.1: 63047801 - 63114267 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01254543_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ino80d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081436.2 NP_001074905.1 5 - 6 1138 57
NM_001114609.1 NP_001108081.1 5 - 6 1229 57
GenBank mRNA AK049468.1 - 5 - 6 1126 57
AK133321.1 - 872 57
BC010584.1 - 5 - 6 1297 57
BC080705.1 - 1 - 2 668 57
BC099533.1 - 1 - 2 656 57
BC150989.1 - 1 - 2 674 9087902 57

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100002442.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
DNA recombination
transcription, DNA-templated
regulation of transcription, DNA-templated
cellular response to DNA damage stimulus
biological_process
molecular_function

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100019870.mm.1 NM_001081436.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100019870.mm.1 NM_001081436.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100019870.mm.1 NM_001114609.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100019870.mm.1 NM_001114609.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100019870.mm.1 TC0100002442.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 740566 - -
ABI Mouse Genome Survey Microarray 827460 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0