This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01254389_g1
See other ARPC2 GE Assays ›
Gene Symbol:
Arpc2
Gene Name:
actin related protein 2/3 complex, subunit 2
Gene Aliases:
2210023N03Rik, 34kDa, p34-Arc
Chromosome Location:
Chr.1: 74236504 - 74268213 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01254389_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arpc2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029711.1 NP_083987.1 5 - 6 738 88
GenBank mRNA AK008777.1 - 1 - 2 184 88
AK083263.1 - 5 - 6 548 88
AK151513.1 - 5 - 6 712 88
AK152356.1 - 6 - 7 563 88
AK170061.1 - 6 - 7 537 88
BC099504.1 - 4 - 5 390 88
BC115750.1 - 5 - 6 536 40051600 88

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of lamellipodium assembly
regulation of actin filament polymerization
positive regulation of actin filament polymerization
Arp2/3 complex-mediated actin nucleation
positive regulation of substrate adhesion-dependent cell spreading
actin binding
structural constituent of cytoskeleton
protein binding
actin filament binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100005000.mm.1 NM_029711.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100005000.mm.1 NM_029711.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100005000.mm.1 TC0100000625.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100005007.mm.1 NM_029711.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100005007.mm.1 NM_029711.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100005007.mm.1 TC0100000625.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100005009.mm.1 NM_029711.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100005009.mm.1 NM_029711.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100005009.mm.1 TC0100000625.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100004999.mm.1 NM_029711.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100004999.mm.1 NM_029711.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100004999.mm.1 TC0100000625.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100005002.mm.1 NM_029711.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100005002.mm.1 NM_029711.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100005002.mm.1 TC0100000625.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100005001.mm.1 NM_029711.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100005001.mm.1 NM_029711.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100005001.mm.1 TC0100000625.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100005006.mm.1 NM_029711.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100005006.mm.1 NM_029711.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100005006.mm.1 TC0100000625.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100005011.mm.1 NM_029711.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100005011.mm.1 NM_029711.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100005011.mm.1 TC0100000625.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100005005.mm.1 NM_029711.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100005005.mm.1 NM_029711.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100005005.mm.1 TC0100000625.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0