This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01254360_g1
See other SLC11A1 GE Assays ›
Gene Symbol:
Slc11a1
Gene Name:
solute carrier family 11 (proton-coupled divalent metal ion transporters), member 1
Gene Aliases:
Bcg, Ity, Lsh, Nramp, Nramp1, ity
Chromosome Location:
Chr.1: 74375014 - 74386057 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01254360_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc11a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013612.2 NP_038640.2 11 - 12 1267 55
GenBank mRNA AK144237.1 - 11 - 12 1252 55
AK171393.1 - 11 - 12 1248 55
BC109137.1 - 11 - 12 1181 40039390 55
BC109138.1 - 11 - 12 1181 40039397 55
L13732.1 - 11 - 12 1245 55
X75355.1 - 11 - 12 1226 55

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc11a1

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

protein import into nucleus, translocation
MAPK cascade
negative regulation of cytokine production
positive regulation of cytokine production
T cell proliferation involved in immune response
T cell cytokine production
positive regulation of dendritic cell antigen processing and presentation
positive regulation of T-helper 1 type immune response
transport
ion transport
iron ion transport
manganese ion transport
cellular cadmium ion homeostasis
cellular iron ion homeostasis
phagocytosis
inflammatory response
vacuolar acidification
response to bacterium
positive regulation of gene expression
nitrite transport
activation of protein kinase activity
response to lipopolysaccharide
interleukin-2 production
interleukin-3 production
positive regulation of interferon-gamma production
response to interferon-gamma
wound healing
macrophage activation
defense response to bacterium
defense response to protozoan
L-arginine import
MHC class II biosynthetic process
respiratory burst
positive regulation of transcription from RNA polymerase II promoter
antigen processing and presentation of peptide antigen
mRNA stabilization
positive regulation of phagocytosis
defense response to Gram-negative bacterium
iron ion homeostasis
multicellular organismal iron ion homeostasis
cadmium ion transmembrane transport
divalent metal ion export
manganese ion transmembrane transport
hydrogen ion transmembrane transport
transporter activity
manganese ion transmembrane transporter activity
protein homodimerization activity
transition metal ion transmembrane transporter activity
metal ion:proton antiporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100005101.mm.1 NM_013612.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100005101.mm.1 NM_013612.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100005101.mm.1 TC0100000632.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0