This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01254092_m1
See other TGFBRAP1 GE Assays ›
Gene Symbol:
Tgfbrap1
Gene Name:
transforming growth factor, beta receptor associated protein 1
Gene Aliases:
3110018K12Rik, AU024090, AU042859, AW742551, Trap1
Chromosome Location:
Chr.1: 43047265 - 43098622 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01254092_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tgfbrap1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013025.2 NP_001013043.1 2 - 3 551 100
GenBank mRNA AK051333.1 - 1 - 2 1786 100
BC072608.1 - 2 - 3 551 100
BC138866.1 - 2 - 3 465 8861888 100
BC138867.1 - 2 - 3 465 8861889 100

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK051333.1, BC138866.1, BC138867.1, NM_001013025.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tgfbrap1

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
intracellular protein transport
signal transduction
vesicle-mediated transport
transforming growth factor beta receptor binding
SMAD binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100018176.mm.1 NM_001013025.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100018176.mm.1 NM_001013025.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100018176.mm.1 TC0100002260.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100018181.mm.1 NM_001013025.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100018181.mm.1 NM_001013025.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100018181.mm.1 TC0100002260.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100018180.mm.1 NM_001013025.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100018180.mm.1 NM_001013025.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100018180.mm.1 TC0100002260.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100018178.mm.1 NM_001013025.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100018178.mm.1 NM_001013025.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100018178.mm.1 TC0100002260.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0