This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01253843_m1
See other SMARCAL1 GE Assays ›
Gene Symbol:
Smarcal1
Gene Name:
SWI/SNF related matrix associated, actin dependent regulator of chromatin, subfamily a-like 1
Gene Aliases:
6030401P21Rik, AI851716, Mharp
Chromosome Location:
Chr.1: 72536687 - 72636790 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01253843_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Smarcal1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018817.2 NP_061287.2 5 - 6 1274 79
GenBank mRNA AF088884.1 - 5 - 6 1254 79
AK042332.1 - 5 - 6 1253 79
AK077878.1 - 5 - 6 1252 79
AK083488.1 - 5 - 6 1252 79
AK146767.1 - 5 - 6 1268 79
AK154020.1 - 5 - 6 1257 79
AK161570.1 - 5 - 6 1194 79
AK169465.1 - 5 - 6 1265 79
BC029078.1 - 5 - 6 1209 3992325 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000608.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Smarcal1

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA strand renaturation
DNA metabolic process
DNA repair
regulation of transcription from RNA polymerase II promoter
cellular response to DNA damage stimulus
replication fork processing
nucleotide binding
DNA binding
helicase activity
ATP binding
DNA-dependent ATPase activity
hydrolase activity
annealing helicase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100004878.mm.1 NM_018817.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100004878.mm.1 NM_018817.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100004878.mm.1 TC0100000608.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100004879.mm.1 NM_018817.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100004879.mm.1 NM_018817.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100004879.mm.1 TC0100000608.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0