This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01253791_g1
See other SLC18A2 GE Assays ›
Gene Symbol:
Slc18a2
Gene Name:
solute carrier family 18 (vesicular monoamine), member 2
Gene Aliases:
1110037L13Rik, 9330105E13, Vmat2
Chromosome Location:
Chr.19: 59260746 - 59296012 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01253791_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc18a2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172523.3 NP_766111.1 7 - 8 950 71
GenBank mRNA AJ555564.1 - 6 - 7 814 71
AK033871.1 - 7 - 8 906 71
AK035644.1 - 7 - 8 933 71
AK041274.1 - 7 - 8 902 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000850.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc18a2

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

response to amphetamine
transport
neurotransmitter transport
serotonin transport
aging
locomotory behavior
response to toxic substance
post-embryonic development
organic cation transport
aminergic neurotransmitter loading into synaptic vesicle
monoamine transport
drug transport
obsolete death
insulin secretion
cellular response to drug
glucose homeostasis
negative regulation of neurotransmitter transport
transmembrane transport
ammonium transmembrane transport
amine transmembrane transporter activity
protein binding
drug binding
serotonin transmembrane transporter activity
drug transmembrane transporter activity
enzyme binding
heat shock protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0