This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01253416_g1
See other CLUH GE Assays ›
Gene Symbol:
Cluh
Gene Name:
clustered mitochondria (cluA/CLU1) homolog
Gene Aliases:
1300001I01Rik, Kiaa0664, mKIAA0664
Chromosome Location:
Chr.11: 74649495 - 74670848 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01253416_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cluh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081158.2 NP_001074627.1 7 - 8 1120 83
GenBank mRNA AB093256.1 - 5 - 6 667 83
AK036540.1 - 7 - 8 1104 83
AK145708.1 - 7 - 8 1162 83
AK150045.1 - 7 - 8 1080 83
AK154146.1 - 7 - 8 1109 83
AK154925.1 - 7 - 8 1121 83
AK157310.1 - 7 - 8 1130 83
BC060069.1 - 2 - 3 232 83
BC072573.1 - 7 - 8 1075 5705116 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001047.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

mitochondrion organization
intracellular distribution of mitochondria
mRNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100009422.mm.1 NM_001081158.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100009422.mm.1 NM_001081158.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100009422.mm.1 TC1100001047.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0