This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01253268_m1
See other HGS GE Assays ›
Gene Symbol:
Hgs
Gene Name:
HGF-regulated tyrosine kinase substrate
Gene Aliases:
Hgr, Hrs, ZFYVE8
Chromosome Location:
Chr.11: 120467635 - 120483984 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01253268_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hgs
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159328.1 NP_001152800.1 8 - 9 760 69
NM_008244.3 NP_032270.3 8 - 9 760 69
GenBank mRNA AK046299.1 - 8 - 9 721 69
AK145031.1 - 8 - 9 680 69
AK166441.1 - 8 - 9 734 69
AK210546.1 - 1 - 2 75 69
AK215004.1 - 1 - 2 137 69
BC003239.1 - 8 - 9 704 3497966 69
D50050.1 - 8 - 9 710 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002028.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hgs

Gene Ontology Categories:

Function(s) Process(es)

protein targeting to lysosome
transport
intracellular protein transport
endosome to lysosome transport
membrane invagination
positive regulation of gene expression
protein transport
regulation of MAP kinase activity
negative regulation of JAK-STAT cascade
protein localization to membrane
positive regulation of exosomal secretion
protein domain specific binding
ubiquitin binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100019507.mm.1 NM_001159328.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100019507.mm.1 NM_001159328.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100019507.mm.1 NM_008244.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100019507.mm.1 NM_008244.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100019507.mm.1 TC1100002028.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0