This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01252060_m1
See other SLC25A19 GE Assays ›
Gene Symbol:
Slc25a19
Gene Name:
solute carrier family 25 (mitochondrial thiamine pyrophosphate carrier), member 19
Gene Aliases:
2900089E13Rik, DNC, MUP1, TPC
Chromosome Location:
Chr.11: 115614170 - 115628295 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01252060_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc25a19
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252384.1 NP_001239313.1 5 - 6 887 94
NM_001252395.1 NP_001239324.1 5 - 6 899 94
NM_001252396.1 NP_001239325.1 3 - 4 554 94
NM_026071.3 NP_080347.1 4 - 5 724 94
GenBank mRNA AK005710.1 - 4 - 5 569 94
AK172622.1 - 3 - 4 526 94
BC018167.1 - 4 - 5 530 3989263 94
BC046767.1 - 4 - 5 574 5694117 94
BI736130.1 - 5 - 6 708 94
CF730205.1 - 3 - 4 365 94

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001252396.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc25a19

Panther Classification:

Molecular Function -

mitochondrial carrier protein

Gene Ontology Categories:

Function(s)

positive regulation of defense response to virus by host
transport
transmembrane transport

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100038412.mm.1 NM_001252384.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038412.mm.1 NM_001252384.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038412.mm.1 NM_001252395.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038412.mm.1 NM_001252395.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038412.mm.1 NM_001252396.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038412.mm.1 NM_001252396.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038412.mm.1 NM_026071.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038412.mm.1 NM_026071.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100038412.mm.1 TC1100004300.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100038411.mm.1 NM_001252384.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038411.mm.1 NM_001252384.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038411.mm.1 NM_001252395.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038411.mm.1 NM_001252395.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038411.mm.1 NM_001252396.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038411.mm.1 NM_001252396.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038411.mm.1 NM_026071.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038411.mm.1 NM_026071.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100038411.mm.1 TC1100004300.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0