This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01251823_g1
See other F2RL3 GE Assays ›
Gene Symbol:
F2rl3
Gene Name:
coagulation factor II (thrombin) receptor-like 3
Gene Aliases:
PAR4
Chromosome Location:
Chr.8: 72761186 - 72763898 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01251823_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: F2rl3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007975.3 NP_032001.2 1 - 2 135 103
GenBank mRNA AF080215.1 - 1 - 2 112 103
AK036427.1 - 1 - 2 136 103
AK153720.1 - 1 - 2 129 103
AK156729.1 - 1 - 2 136 103

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene F2rl3

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
blood coagulation
hemostasis
positive regulation of release of sequestered calcium ion into cytosol
thrombin receptor signaling pathway
signal transducer activity
G-protein coupled receptor activity
thrombin receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0