This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01251624_g1
See other GCDH GE Assays ›
Gene Symbol:
Gcdh
Gene Name:
glutaryl-Coenzyme A dehydrogenase
Gene Aliases:
9030411L18, AI266902, D17825, GCD
Chromosome Location:
Chr.8: 84886387 - 84893921 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01251624_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gcdh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001044744.1 NP_001038209.2 9 - 10 1160 102
NM_008097.2 NP_032123.3 8 - 9 1217 102
GenBank mRNA AK078866.1 - 6 - 7 770 102
AK079500.1 - 2 - 3 268 102
AK165406.1 - 8 - 9 1043 102
BC061158.1 - 8 - 9 1047 30314273 102
U18992.1 - 8 - 9 998 102

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gcdh

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

acyl-CoA metabolic process
metabolic process
fatty acid oxidation
fatty-acyl-CoA biosynthetic process
oxidation-reduction process
fatty-acyl-CoA binding
acyl-CoA dehydrogenase activity
glutaryl-CoA dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-CH group of donors
flavin adenine dinucleotide binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0