This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01251294_g1
See other EIF2S2 GE Assays ›
Gene Symbol:
Eif2s2
Gene Name:
eukaryotic translation initiation factor 2, subunit 2 (beta)
Gene Aliases:
2810026E11Rik, 38kDa, AA408636, AA571381, AA986487, AW822225, D2Ertd303e, EIF2, EIF2B
Chromosome Location:
Chr.2: 154871410 - 154892906 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01251294_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif2s2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026030.2 NP_080306.1 1 - 2 208 116
GenBank mRNA AK011503.1 - 1 - 2 129 116
AK012338.1 - 1 - 2 121 116
AK076056.1 - 1 - 2 128 116
AK143553.1 - 1 - 2 116
AK145401.1 - 1 - 2 113 116
AK145434.1 - 1 - 2 155 116
AK166862.1 - 1 - 2 112 116
AK166866.1 - 1 - 2 136 116
BC003848.1 - 1 - 2 84 3594058 116
BC100539.1 - 1 - 2 119 116
BC113766.1 - 1 - 2 80 40058503 116
CJ084276.1 - 1 - 2 208 116

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK011503.1,AK145401.1,AK145434.1,AK166862.1,BC003848.1,BC113766.1,NM_026030.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif2s2

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
male germ cell proliferation
translation
translational initiation
male gonad development
translation initiation factor activity
protein binding
poly(A) RNA binding
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200042000.mm.1 NM_026030.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200042000.mm.1 NM_026030.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200042000.mm.1 TC0200004951.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200041989.mm.1 NM_026030.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200041989.mm.1 NM_026030.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200041989.mm.1 TC0200004951.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200041994.mm.1 NM_026030.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200041994.mm.1 NM_026030.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200041994.mm.1 TC0200004951.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 383544 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0