This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01251288_m1
See other MYLK2 GE Assays ›
Gene Symbol:
Mylk2
Gene Name:
myosin, light polypeptide kinase 2, skeletal muscle
Gene Aliases:
9830004H17Rik, BB138278
Chromosome Location:
Chr.2: 152911276 - 152923065 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01251288_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mylk2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081044.2 NP_001074513.2 5 - 6 1218 62
GenBank mRNA BC158041.1 - 5 - 6 1049 9088157 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002303.mm.1

Set Membership:

Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
water transport
regulation of muscle contraction
striated muscle contraction
neuromuscular synaptic transmission
positive regulation of gene expression
skeletal muscle satellite cell differentiation
phosphorylation
peptidyl-threonine phosphorylation
positive regulation of fast-twitch skeletal muscle fiber contraction
regulation of muscle filament sliding
skeletal muscle cell differentiation
regulation of MAPK cascade
protein autophosphorylation
regulation of neuronal synaptic plasticity
synaptic vesicle transport
regulation of cell cycle
cardiac muscle cell differentiation
cardiac muscle tissue morphogenesis
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
myosin light chain kinase activity
protein binding
calmodulin binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
myosin light chain binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200018028.mm.1 NM_001081044.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200018028.mm.1 NM_001081044.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200018028.mm.1 TC0200002303.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0