This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01251230_m1
See other ATMIN GE Assays ›
Gene Symbol:
Atmin
Gene Name:
ATM interactor
Gene Aliases:
Asciz, gpg6, mKIAA0431
Chromosome Location:
Chr.8: 116943386 - 116960445 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01251230_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atmin
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177700.4 NP_808368.3 2 - 3 486 70
GenBank mRNA AK122281.1 - 2 - 3 368 70
AX775769.1 - 2 - 3 486 70
BC027752.1 - 2 - 3 460 70
BC060631.1 - 2 - 3 471 6847850 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001392.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atmin

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
cellular response to DNA damage stimulus
positive regulation of transcription, DNA-templated
cilium morphogenesis
transcription regulatory region DNA binding
dynein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800011317.mm.1 NM_177700.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011317.mm.1 NM_177700.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800011317.mm.1 TC0800001392.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800011318.mm.1 NM_177700.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011318.mm.1 NM_177700.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800011318.mm.1 TC0800001392.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0