This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01251073_m1
See other ATP6V0D1 GE Assays ›
Gene Symbol:
Atp6v0d1
Gene Name:
ATPase, H+ transporting, lysosomal V0 subunit D1
Gene Aliases:
AI267038, Ac39, Atp6d, P39, VATX, Vma6
Chromosome Location:
Chr.8: 105524470 - 105566040 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01251073_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp6v0d1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013477.3 NP_038505.2 2 - 3 420 91
GenBank mRNA AB088408.1 - 2 - 3 309 91
AK083372.1 - 2 - 3 421 91
AK133921.1 - 2 - 3 428 91
AK171515.1 - 2 - 3 390 91
BC011075.1 - 2 - 3 311 3662404 91
U13840.1 - 2 - 3 309 91
U21549.1 - 2 - 3 292 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800002824.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp6v0d1

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
ATP hydrolysis coupled proton transport
proton transport
cell projection organization
hydrogen ion transmembrane transport
hydrogen-exporting ATPase activity, phosphorylative mechanism
hydrogen ion transmembrane transporter activity
protein complex binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800021748.mm.1 NM_013477.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800021748.mm.1 NM_013477.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800021748.mm.1 TC0800002824.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0