This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01251059_g1
See other SLC9A5 GE Assays ›
Gene Symbol:
Slc9a5
Gene Name:
solute carrier family 9 (sodium/hydrogen exchanger), member 5
Gene Aliases:
Gm696
Chromosome Location:
Chr.8: 105348104 - 105369881 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01251059_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc9a5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081332.1 NP_001074801.1 5 - 6 2038 79
GenBank mRNA AK134092.1 - 1 - 2 133 79
BC057616.1 - 2 - 3 574 79
BC151197.1 - 5 - 6 1479 9088085 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001215.mm.1

Set Membership:

Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

sodium ion transport
proton transport
sodium ion transmembrane transport
hydrogen ion transmembrane transport
sodium:proton antiporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800009594.mm.1 NM_001081332.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800009594.mm.1 NM_001081332.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800009594.mm.1 TC0800001215.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0