Assay Details
Gene Symbol: Slc9a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081332.1 | NP_001074801.1 | 11 - 12 | 2844 | 97 | |
GenBank mRNA | AK134092.1 | - | 5 - 6 | 1087 | 97 | |
BC057616.1 | - | 6 - 7 | 1528 | 97 | ||
BC151197.1 | - | 11 - 12 | 2285 | 9088085 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
