This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01249786_g1
See other AURKA GE Assays ›
Gene Symbol:
Aurka
Gene Name:
aurora kinase A
Gene Aliases:
AIRK1, ARK-1, AU019385, AW539821, Ark1, Aurora-A, Ayk1, IAK, IAK1, Stk6
Chromosome Location:
Chr.2: 172356190 - 172370706 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01249786_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aurka
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001291185.1 NP_001278114.1 2 - 3 139 95
GenBank mRNA AK077513.1 - 2 - 3 95 95
AK085861.1 - 2 - 3 91 95
AK145968.1 - 2 - 3 105 95
AK169363.1 - 2 - 3 94 95
BC005425.1 - 2 - 3 84 3590706 95
U69106.1 - 2 - 3 75 95
U80932.1 - 2 - 3 84 95

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK077513.1,AK085861.1,AK145968.1,AK169363.1,BC005425.1,NM_001291185.1,U69106.1,U80932.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

meiotic spindle organization
microtubule cytoskeleton organization
mitotic cell cycle
protein phosphorylation
cell cycle
spindle organization
mitotic spindle organization
spindle assembly involved in female meiosis I
mitotic nuclear division
mitotic centrosome separation
meiotic nuclear division
anterior/posterior axis specification
phosphorylation
cell projection organization
regulation of protein stability
negative regulation of protein binding
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
regulation of cytokinesis
histone-serine phosphorylation
negative regulation of apoptotic process
centrosome organization
cell division
centrosome localization
protein localization to centrosome
negative regulation of spindle checkpoint
positive regulation of oocyte maturation
neuron projection extension
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
protein serine/threonine/tyrosine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein kinase binding
ubiquitin protein ligase binding
histone serine kinase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200045000.mm.1 NM_011497.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200045000.mm.1 TC0200005271.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200045002.mm.1 NM_011497.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200045002.mm.1 TC0200005271.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200044999.mm.1 NM_011497.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200044999.mm.1 TC0200005271.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0