This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01249743_m1
See other ZNFX1 GE Assays ›
Gene Symbol:
Znfx1
Gene Name:
zinc finger, NFX1-type containing 1
Gene Aliases:
AI481105, Astx3
Chromosome Location:
Chr.2: 167035790 - 167063015 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01249743_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Znfx1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033196.2 NP_001028368.2 4 - 5 2540 85
GenBank mRNA AK147564.1 - 4 - 5 2542 85
AK147652.1 - 4 - 5 2542 85
AK149249.1 - 4 - 5 2159 85
AK150223.1 - 4 - 5 2129 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200005181.mm.1

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
poly(A) RNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200044345.mm.1 NM_001033196.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200044345.mm.1 NM_001033196.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200044345.mm.1 TC0200005181.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0