This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01249476_m1
See other C330019F10RIK GE Assays ›
Gene Symbol:
C330019F10Rik
Gene Name:
RIKEN cDNA C330019F10 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01249476_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C330019F10Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK049282.1 - 4 - 5 336 65
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB400623.2 - 4 - 5 373 Chr. 11 - 107511401 - 107527223 [-] on Build GRCm38 65
GenBankmRNA BB651899.1 - 4 - 5 336 Chr. 11 - 107511400 - 107527220 [-] on Build GRCm38 65

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream proteasome (prosome,
macropain) 26S subunit,
non-ATPase,
12
Psmd12 1500002F15Rik,
AI480719,
P55
-13.4kb Anti-sense Chr. 11 - 107479528 - 107498036 on NCBI Build GRCm38
Downstream RIKEN cDNA A830035A12 gene A830035A12Rik 2.0kb Sense Chr. 11 - 107526506 - 107534143 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

MicroArray Probes:

Agilent

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 418184 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 952293 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 504333 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 684588 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 383002 AK042452.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 383002 BC051092.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 383002 NM_009475.1 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 383002 U28486.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 743137 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 555556 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 673006 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 111900 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106968 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106968 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07482655 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123726 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106968 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107504 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107506 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107509 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107496 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107483 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107501 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107505 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107456 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09150280 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107521 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09671220 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107526 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107503 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107441 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107444 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09329103 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107446 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107484 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107518 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107466 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107486 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107525 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107493 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107458 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107455 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107454 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107511 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107461 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09229871 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107524 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09482300 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107452 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09119731 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107465 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09686574 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107517 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107459 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107463 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107468 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09516327 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107450 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09437202 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107523 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107513 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28107498 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0