This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01249474_g1
See other C330019F10RIK GE Assays ›
Gene Symbol:
C330019F10Rik
Gene Name:
RIKEN cDNA C330019F10 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01249474_g1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C330019F10Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK049282.1 - 1 - 2 48 84
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB400623.2 - 1 - 2 85 Chr. 11 - 107511401 - 107527223 [-] on Build GRCm38 84
GenBankmRNA BB651899.1 - 1 - 2 48 Chr. 11 - 107511400 - 107527220 [-] on Build GRCm38 84

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RIKEN cDNA A830035A12 gene A830035A12Rik Sense Chr. 11 - 107526506 - 107534143 on NCBI Build GRCm38
Downstream helicase with zinc finger domain Helz 3110078M01Rik,
9430093I07Rik,
9630002H22Rik,
AI851979,
DRHC
19.7kb Anti-sense Chr. 11 - 107546958 - 107693826 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus 1700 Probe spans exons

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 464957 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0