This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01249275_g1
See other SIN3B GE Assays ›
Gene Symbol:
Sin3b
Gene Name:
transcriptional regulator, SIN3B (yeast)
Gene Aliases:
2810430C10Rik
Chromosome Location:
Chr.8: 72723270 - 72758203 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01249275_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sin3b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009188.4 NP_033214.2 9 - 10 1275 79
GenBank mRNA AK087970.1 - 2 - 3 372 79
AK159076.1 - 9 - 10 1258 79
AK173005.1 - 8 - 9 3225 79
BC129859.1 - 9 - 10 1247 79
L38622.2 - 9 - 10 1247 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800000834.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sin3b

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
skeletal muscle tissue development
histone deacetylation
negative regulation of cell cycle
negative regulation of transcription, DNA-templated
cardiac muscle tissue development
RNA polymerase II transcription corepressor activity
chromatin binding
transcription corepressor activity
histone deacetylase activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800006179.mm.1 NM_009188.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800006179.mm.1 TC0800000834.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0