This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01249213_g1
See other CDH3 GE Assays ›
Gene Symbol:
Cdh3
Gene Name:
cadherin 3
Gene Aliases:
AI385538, Cadp, Cdhp, P-cadherin, Pcad
Chromosome Location:
Chr.8: 106510852 - 106556911 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01249213_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdh3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037809.5 NP_001032898.1 4 - 5 476 92
NM_007665.3 NP_031691.1 4 - 5 473 92
GenBank mRNA AK031265.1 - 4 - 5 416 92
AK045041.1 - 4 - 5 436 92
BC052189.1 - 2 - 3 164 92
BC098459.1 - 4 - 5 382 30079668 92
X06340.1 - 4 - 5 414 92

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdh3

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

retina homeostasis
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
positive regulation of gene expression
positive regulation of keratinocyte proliferation
single organismal cell-cell adhesion
hair cycle process
keratinization
positive regulation of monophenol monooxygenase activity
negative regulation of transforming growth factor beta2 production
positive regulation of insulin-like growth factor receptor signaling pathway
positive regulation of melanin biosynthetic process
negative regulation of catagen
canonical Wnt signaling pathway
regulation of hair cycle by canonical Wnt signaling pathway
positive regulation of melanosome transport
calcium ion binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0