This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01249022_g1
See other GM14169 GE Assays ›
Gene Symbol:
Gm14169
Gene Name:
predicted gene 14169
Gene Aliases:
OTTMUSG00000015869
Chromosome Location:
Chr.2: 156609198 - 156613422 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01249022_g1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gm14169
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_040372.1 2 - 3 393 93
GenBank mRNA AK032272.1 - 2 - 3 394 93
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB622598.1 - 2 - 3 394 Chr. 2 - 156611071 - 156613422 [-] on Build GRCm38 93

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream AAR2 splicing factor homolog (S. cerevisiae) Aar2 0610011L14Rik -43.5kb Anti-sense Chr. 2 - 156547558 - 156568972 on NCBI Build GRCm38
Downstream discs,
large homolog-associated protein 4 (Drosophila)
Dlgap4 AI225853,
BC024558,
DAP-4,
DAP4,
SAPAP-4,
Sapap4,
WBP16
437.0 Anti-sense Chr. 2 - 156613633 - 156764363 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200004994.mm.1

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200042672.mm.1 NR_040372.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200042672.mm.1 NR_040372.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200042672.mm.1 TC0200004994.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0