This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01248117_m1
See other EYA2 GE Assays ›
Gene Symbol:
Eya2
Gene Name:
eyes absent 2 homolog (Drosophila)
Gene Aliases:
Eab1
Chromosome Location:
Chr.2: 165595028 - 165771727 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01248117_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eya2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271962.1 NP_001258891.1 8 - 9 856 81
NM_001271963.1 NP_001258892.1 8 - 9 1032 81
NM_010165.3 NP_034295.1 8 - 9 1033 81
GenBank mRNA AK137718.1 - 8 - 9 1028 81
AK141765.1 - 8 - 9 1032 81
AY212511.1 - 5 - 6 547 81
AY212512.1 - 7 - 8 793 81
BC003755.1 - 8 - 9 1033 3497245 81
U61111.1 - 7 - 8 774 81
U71208.1 - 8 - 9 858 81
U81603.1 - 7 - 8 773 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002550.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eya2

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
transcription, DNA-templated
regulation of transcription, DNA-templated
protein dephosphorylation
cellular response to DNA damage stimulus
multicellular organism development
striated muscle tissue development
chromatin modification
histone dephosphorylation
peptidyl-tyrosine dephosphorylation
extrinsic apoptotic signaling pathway in absence of ligand
mitochondrial outer membrane permeabilization
magnesium ion binding
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
protein binding
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200020960.mm.1 NM_001271962.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200020960.mm.1 NM_001271963.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200020960.mm.1 NM_010165.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200020960.mm.1 TC0200002550.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0