Assay Details
Gene Symbol: Usb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133954.2 | NP_598715.2 | 6 - 7 | 772 | 76 | |
GenBank mRNA | AK135527.1 | - | 6 - 7 | 765 | 76 | |
AK160901.1 | - | 6 - 7 | 755 | 76 | ||
AK168683.1 | - | 6 - 7 | 754 | 76 | ||
BC016418.1 | - | 6 - 7 | 716 | 4224590 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
mRNA processing
RNA splicing
U6 snRNA 3'-end processing
RNA phosphodiester bond hydrolysis, exonucleolytic
3'-5'-exoribonuclease activity
cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
calmodulin-dependent cyclic-nucleotide phosphodiesterase activity
cGMP-stimulated cyclic-nucleotide phosphodiesterase activity
cGMP-inhibited cyclic-nucleotide phosphodiesterase activity
photoreceptor cyclic-nucleotide phosphodiesterase activity
nuclease activity
hydrolase activity
7,8-dihydro-D-neopterin 2',3'-cyclic phosphate phosphodiesterase activity
inositol phosphosphingolipid phospholipase activity
inositol phosphorylceramide phospholipase activity
mannosyl-inositol phosphorylceramide phospholipase activity
mannosyl-diinositol phosphorylceramide phospholipase activity
mRNA processing
RNA splicing
U6 snRNA 3'-end processing
RNA phosphodiester bond hydrolysis, exonucleolytic
3'-5'-exoribonuclease activity
cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
calmodulin-dependent cyclic-nucleotide phosphodiesterase activity
cGMP-stimulated cyclic-nucleotide phosphodiesterase activity
cGMP-inhibited cyclic-nucleotide phosphodiesterase activity
photoreceptor cyclic-nucleotide phosphodiesterase activity
nuclease activity
hydrolase activity
7,8-dihydro-D-neopterin 2',3'-cyclic phosphate phosphodiesterase activity
inositol phosphosphingolipid phospholipase activity
inositol phosphorylceramide phospholipase activity
mannosyl-inositol phosphorylceramide phospholipase activity
mannosyl-diinositol phosphorylceramide phospholipase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26082719 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26082717 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26082718 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P212977 | AK168683.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P212977 | BC016418.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P212977 | NM_133954.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P212977 | AK135527.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P212977 | AK160901.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P212977 | AK168683.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P212977 | BC016418.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P212977 | NM_133954.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P212977 | AK135527.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P212977 | AK160901.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 353761 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731770 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 259409 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608154 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 468070 | XM_001472087.1 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 468070 | XM_001476334.1 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 468070 | XM_001478015.1 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 721787 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 316434 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 651454 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 933509 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 138612 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154948 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P131187 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P212977 | AK168683.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P212977 | BC016418.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P212977 | NM_133954.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P212977 | AK135527.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P212977 | AK160901.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P212977 | AK168683.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P212977 | BC016418.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P212977 | NM_133954.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P212977 | AK135527.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P212977 | AK160901.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26082721 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26082725 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26082794 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26082727 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26082724 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26082718 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26082723 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26082733 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26082730 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26082791 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26082793 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154948 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06605119 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P131187 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01802794 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06605098 | - | - |
Whole Mouse Genome G4122A | A_51_P212977 | AK168683.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P212977 | BC016418.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P212977 | NM_133954.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P212977 | AK135527.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P212977 | AK160901.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P212977 | AK168683.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P212977 | BC016418.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P212977 | NM_133954.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P212977 | AK135527.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P212977 | AK160901.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132472 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120979 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P131187 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154948 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07169042 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07300141 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26082717 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26082718 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26082719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082820 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082794 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082862 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082724 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082839 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082783 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082845 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082859 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082868 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082866 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082842 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082863 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082721 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082822 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082827 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082713 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082869 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082813 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082818 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082840 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082838 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082854 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082796 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082844 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082830 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082861 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082852 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082815 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082847 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082825 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082849 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082851 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082803 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082718 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082781 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082769 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082723 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082731 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082743 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06673775 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082789 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082725 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082787 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07168078 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082737 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07169044 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082785 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082776 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082729 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06877250 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082791 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082735 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082793 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082794 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082744 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082734 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07032921 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082772 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082783 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082736 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082770 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07050004 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07300141 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082727 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06877248 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082724 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082779 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082741 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10574345 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574346 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574349 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574347 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574344 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574343 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1448583_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10574342 | - | - |
Mouse Genome 430 2.0 Array | 1448583_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1448583_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 101985_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 101985_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0800008928.mm.1 | NM_133954.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800008928.mm.1 | NM_133954.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800008928.mm.1 | TC0800001132.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800008927.mm.1 | NM_133954.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800008927.mm.1 | NM_133954.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800008927.mm.1 | TC0800001132.mm.1 | MTA TC |
Mouse Expression Set 430 | 1448583_at | - | - |
Mouse Expression Set 430 | 1448583_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1448583_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1448583_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448583_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448583_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 534721 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 1400017 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001400017 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2741445 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2741445 | - | - |
mouse-6 v1.1 (Illumina) | scl33463.8.1_0-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1400017 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl33463.8.1_0-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1400017 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2741445 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3008733 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2741445 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3008733 | - | - |
