This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01247230_m1
See other INPP4B GE Assays ›
Gene Symbol:
Inpp4b
Gene Name:
inositol polyphosphate-4-phosphatase, type II
Gene Aliases:
E130107I17Rik
Chromosome Location:
Chr.8: 81342342 - 82152019 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01247230_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Inpp4b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001024617.3 NP_001019788.1 8 - 9 928 69
NM_001297591.1 NP_001284520.1 8 - 9 905 69
NM_001297593.1 NP_001284522.1 7 - 8 786 69
NM_001297596.1 NP_001284525.1 8 - 9 905 69
GenBank mRNA AK053554.1 - 1 - 2 57 69
AK089068.1 - 3 - 4 159 69
AK139397.1 - 1 - 2 406 69
AK141502.1 - 10 - 11 902 69
BC064813.1 - 6 - 7 630 6826283 69
DQ296611.1 - 8 - 9 905 69
DQ296612.1 - 7 - 8 786 69
DQ296613.1 - 8 - 9 905 69

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Inpp4b

Gene Ontology Categories:

Function(s) Process(es)

cellular calcium ion homeostasis
dephosphorylation
phosphatidylinositol-3-phosphate biosynthetic process
negative regulation of osteoclast differentiation
regulation of nucleocytoplasmic transport
regulation of bone remodeling
inositol phosphate dephosphorylation
phosphatidylinositol dephosphorylation
regulation of protein kinase B signaling
lipid binding
phosphatidylinositol-3,4-bisphosphate 4-phosphatase activity
hydrolase activity
phosphatidylinositol bisphosphate phosphatase activity
phosphatidylinositol trisphosphate phosphatase activity
phosphatidylinositol-4,5-bisphosphate 4-phosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0