This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01247167_m1
See other MGAT4D GE Assays ›
Gene Symbol:
Mgat4d
Gene Name:
MGAT4 family, member C
Gene Aliases:
4933434I20Rik, GnT1IP, GnT1IP-L
Chromosome Location:
Chr.8: 83339402 - 83382299 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01247167_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mgat4d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026233.2 NP_080509.2 4 - 5 537 101
GenBank mRNA AK016272.1 - 4 - 5 546 101
AK017057.1 - 1 - 2 127 101
HM067443.1 - 4 - 5 661 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800000921.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mgat4d

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
biological_process
alpha-1,6-mannosyltransferase activity
alpha-1,2-mannosyltransferase activity
mannosyltransferase activity
alpha-1,3-mannosyltransferase activity
alpha-1,3-galactosyltransferase activity
molecular_function
UDP-glucose:glycoprotein glucosyltransferase activity
glycolipid mannosyltransferase activity
oligosaccharyl transferase activity
dolichyl-phosphate-glucose-glycolipid alpha-glucosyltransferase activity
acetylglucosaminyltransferase activity
acetylgalactosaminyltransferase activity
galactosyltransferase activity
fucosyltransferase activity
O antigen polymerase activity
lipopolysaccharide-1,6-galactosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
transferase activity, transferring hexosyl groups
cellulose synthase activity
9-phenanthrol UDP-glucuronosyltransferase activity
1-phenanthrol glycosyltransferase activity
9-phenanthrol glycosyltransferase activity
1,2-dihydroxy-phenanthrene glycosyltransferase activity
phenanthrol glycosyltransferase activity
beta-1,4-mannosyltransferase activity
alpha-1,2-galactosyltransferase activity
dolichyl pyrophosphate Man7GlcNAc2 alpha-1,3-glucosyltransferase activity
lipopolysaccharide-1,5-galactosyltransferase activity
dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
inositol phosphoceramide synthase activity
glucosyltransferase activity
alpha-(1->3)-fucosyltransferase activity
alpha-(1->6)-fucosyltransferase activity
indole-3-butyrate beta-glucosyltransferase activity
salicylic acid glucosyltransferase (ester-forming) activity
salicylic acid glucosyltransferase (glucoside-forming) activity
benzoic acid glucosyltransferase activity
chondroitin hydrolase activity
dolichyl-pyrophosphate Man7GlcNAc2 alpha-1,6-mannosyltransferase activity
cytokinin 9-beta-glucosyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800006796.mm.1 NM_026233.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006796.mm.1 NM_026233.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800006796.mm.1 TC0800000921.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800006800.mm.1 NM_026233.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006800.mm.1 NM_026233.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800006800.mm.1 TC0800000921.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800006797.mm.1 NM_026233.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006797.mm.1 NM_026233.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800006797.mm.1 TC0800000921.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800006798.mm.1 NM_026233.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006798.mm.1 NM_026233.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800006798.mm.1 TC0800000921.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800006801.mm.1 NM_026233.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006801.mm.1 NM_026233.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800006801.mm.1 TC0800000921.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0