This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01247128_g1
See other ANAPC10 GE Assays ›
Gene Symbol:
Anapc10
Gene Name:
anaphase promoting complex subunit 10
Gene Aliases:
1500026N15Rik, A830003M23Rik, APC10
Chromosome Location:
Chr.8: 79711755 - 79777321 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01247128_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Anapc10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026904.2 NP_081180.1 2 - 3 198 93
GenBank mRNA AK041373.1 - 250 93
AK219569.1 - 2 - 3 193 93
BC031460.1 - 1 - 2 198 3710036 93

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Anapc10

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
mitotic nuclear division
regulation of mitotic metaphase/anaphase transition
anaphase-promoting complex-dependent catabolic process
cell division
protein K11-linked ubiquitination
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0