This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01247005_g1
See other EPB4.1L1 GE Assays ›
Gene Symbol:
Epb4.1l1
Gene Name:
erythrocyte protein band 4.1 like 1
Gene Aliases:
4.1N, Epb41l1, NBL1, mKIAA0338
Chromosome Location:
Chr.2: 156420487 - 156543214 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01247005_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Epb4.1l1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001006664.3 NP_001006665.1 20 - 21 3003 84
NM_001291120.1 NP_001278049.1 21 - 22 2970 84
NM_001291121.1 NP_001278050.1 20 - 21 2934 84
NM_001291122.1 NP_001278051.1 19 - 20 2526 84
NM_001291123.1 NP_001278052.1 19 - 20 2523 84
NM_013510.4 NP_038538.1 20 - 21 2921 84
GenBank mRNA AB370257.1 - 2 - 3 164 84
AB370262.1 - 19 - 20 2516 84
AF061283.1 - 20 - 21 2735 84
AK122251.1 - 20 - 21 2601 84
AK147272.1 - 19 - 20 2385 84
AK147611.1 - 21 - 22 2972 84
AK154704.1 - 19 - 20 2345 84
BC034751.1 - 20 - 21 2731 4954660 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002395.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Epb4.1l1

Gene Ontology Categories:

Function(s) Process(es)

cortical actin cytoskeleton organization
actin binding
receptor binding
structural molecule activity
protein binding
cytoskeletal protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200019295.mm.1 NM_001006664.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200019295.mm.1 NM_013510.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200019295.mm.1 TC0200002395.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0