This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01246373_g1
See other MRPL38 GE Assays ›
Gene Symbol:
Mrpl38
Gene Name:
mitochondrial ribosomal protein L38
Gene Aliases:
1110036N21Rik, 4733401F03Rik, MRP-L3, R75240, Rpml3
Chromosome Location:
Chr.11: 116131817 - 116138868 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01246373_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mrpl38
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024177.3 NP_077139.2 3 - 4 476 90
GenBank mRNA AK088343.1 - 3 - 4 415 90
AK151598.1 - 3 - 4 415 90
AK159033.1 - 3 - 4 477 90
BC002319.1 - 2 - 3 452 3593984 90
BC030739.1 - 3 - 4 403 1493919 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100004104.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mrpl38

Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100038708.mm.1 NM_024177.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038708.mm.1 NM_024177.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100038708.mm.1 TC1100004104.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0