This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01246353_g1
See other UBE2O GE Assays ›
Gene Symbol:
Ube2o
Gene Name:
ubiquitin-conjugating enzyme E2O
Gene Aliases:
9630022H21, B230113M03Rik, E2-230K, mKIAA1734
Chromosome Location:
Chr.11: 116537740 - 116581476 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01246353_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ube2o
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_173755.3 NP_776116.2 6 - 7 915 76
GenBank mRNA AK129431.1 - 6 - 7 915 76
BC059193.1 - 6 - 7 540 4160667 76
U20780.1 - 6 - 7 577 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100004121.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ube2o

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
protein monoubiquitination
positive regulation of BMP signaling pathway
retrograde transport, endosome to Golgi
protein K63-linked ubiquitination
nucleotide binding
ubiquitin-protein transferase activity
ATP binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
ubiquitin protein ligase binding
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
poly(A) RNA binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100039038.mm.1 NM_173755.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100039038.mm.1 NM_173755.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100039038.mm.1 TC1100004121.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0