This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01245924_m1
See other SLC4A1 GE Assays ›
Gene Symbol:
Slc4a1
Gene Name:
solute carrier family 4 (anion exchanger), member 1
Gene Aliases:
Ae1, CD233, Empb3, l11Jus51
Chromosome Location:
Chr.11: 102348820 - 102365281 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01245924_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc4a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011403.2 NP_035533.1 17 - 18 2658 78
GenBank mRNA AK137542.1 - 17 - 18 2554 78
AK157181.1 - 17 - 18 2627 78
AK157833.1 - 17 - 18 2516 78
AK158243.1 - 17 - 18 2528 78
AK158453.1 - 17 - 18 2503 78
AK162616.1 - 17 - 18 2503 78
AK165195.1 - 17 - 18 2515 78
AK165267.1 - 17 - 18 2522 78
AK171662.1 - 2554 78
AK171808.1 - 17 - 18 2492 78
AK171845.1 - 2554 78
AK172103.1 - 17 - 18 2515 78
AK172316.1 - 17 - 18 2503 78
AK172395.1 - 17 - 18 2554 78
AK172456.1 - 17 - 18 2538 78
AK172569.1 - 17 - 18 2503 78
AY296129.1 - 17 - 18 2491 78
BC052419.1 - 17 - 18 2498 5702599 78
BC053429.1 - 17 - 18 2489 6506270 78
M29379.1 - 17 - 18 2485 78
X02677.1 - 17 - 18 2491 78
X03917.1 - 15 - 16 2335 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003833.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc4a1

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
anion transport
chloride transport
inorganic anion transport
bicarbonate transport
regulation of intracellular pH
anion transmembrane transport
chloride transmembrane transport
actin binding
transporter activity
inorganic anion exchanger activity
protein binding
protein C-terminus binding
anion transmembrane transporter activity
bicarbonate transmembrane transporter activity
chloride transmembrane transporter activity
anion:anion antiporter activity
enzyme binding
ankyrin binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100035657.mm.1 NM_011403.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100035657.mm.1 NM_011403.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100035657.mm.1 TC1100003833.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0